MilliporeSigma
All Photos(2)

243620

Sigma-Aldrich

Sodium borohydride on aluminum oxide

extent of labeling: ~10 wt. % loading, matrix basic alumina support

Linear Formula:
NaBH4 · (Al2O3)x
MDL number:
PubChem Substance ID:

form

powder

Quality Level

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: carbonyl reactive

extent of labeling

~10 wt. % loading

matrix

basic alumina support

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Packaging

10, 50 g in glass bottle

Application

Supported reagent which is often more selective and easier to use than free NaBH4.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B - Skin Corr. 1B - Water-react. 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service