Merck
Multimedia Durapore® 0.2/0.22 µm, Opticap® XL Capsule Opticap® XL 10, inlet connection diam. 9/16 in., pore size 0.2/0.22 μm, cartridge nominal length 10 in. (25 cm)
Multimedia Durapore® 0.2/0.22 µm, Opticap® XL Capsule
Multimedia Durapore® 0.2/0.22 µm, Opticap® XL Capsule Opticap® XL 10, inlet connection diam. 9/16 in., pore size 0.2/0.22 μm, cartridge nominal length 10 in. (25 cm)