HPLC Analysis of Prednisolone (Secondary Pharma Standard) on Ascentis® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis C18, 15 cm x 4.6 mm I.D., 5 μm particles (581324-U)
mobile phase
[A] water; [B] methanol:acetonitrile (50:50, A:B)
gradient
40% B for 14 min; to 80% B in 6 min; held at 80% B for 5 min
flow rate
1 mL/min
column temp.
40 °C
detector
UV, 254 nm
injection
10 μL
sample
125 μg/mL in water:acetonitrile (60:40)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany