Merck
  • Local Lattice Distortion Activate Metastable Metal Sulfide as Catalyst with Stable Full Discharge-Charge Capability for Li-O
Local Lattice Distortion Activate Metastable Metal Sulfide as Catalyst with Stable Full Discharge-Charge Capability for Li-O
Nano letters (2017-05-10)
Sanpei Zhang, Zhennan Huang, Zhaoyin Wen, Linlin Zhang, Jun Jin, Reza Shahbazian-Yassar, Jianhua Yang
ABSTRACT
The direct lattice strain, either distortion, compressive, or tensile, can efficiently alter the intrinsic electrocatalytic property of the catalysts. In this work, we report a novel and effective strategy to distort the lattice structure by constructing a metastable MoSSe solid solution and thus, tune its catalytic activity for the Li-O
Sigma-Aldrich
Selenourea, 98%