Merck
  • Protopine reduces the inflammatory activity of lipopolysaccharide-stimulated murine macrophages.
Protopine reduces the inflammatory activity of lipopolysaccharide-stimulated murine macrophages.
BMB reports (2012-03-01)
Deok Sung Bae, Young Hoon Kim, Cheol-Ho Pan, Chu Won Nho, Javzan Samdan, Jamyansan Yansan, Jae Kown Lee
ABSTRACT
Protopine is an isoquinoline alkaloid contained in plants in northeast Asia. In this study, we investigated whether protopine derived from Hypecoum erectum L could suppress lipopolysaccharide (LPS)-induced inflammatory responses in murine macrophages (Raw 264.7 cells). Protopine was found to reduce nitric oxide (NO), cyclooxygenase-2 (COX-2), and prostaglandin E(2) (PGE(2)) production by LPS-stimulated Raw 264.7 cells, without a cytotoxic effect. Pre-treatment of Raw 264.7 cells with protopine reduced the production of pro-inflammatory cytokines. These inhibitory effects were caused by blocking phosphorylation of mitogen-activated protein kinases (MAP kinases) and also blocking activation of a nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB).
Sigma-Aldrich
Protopine hydrochloride, ≥98%, solid