Merck
  • Identification of an MSI-H tumor-specific cytotoxic T cell epitope generated by the (-1) frame of U79260(FTO).
Identification of an MSI-H tumor-specific cytotoxic T cell epitope generated by the (-1) frame of U79260(FTO).
Journal of biomedicine & biotechnology (2010-03-27)
Michael Linnebacher, Anne Wienck, Inga Boeck, Ernst Klar
ABSTRACT
Microsatellite instability (MSI-H) induced by defects of the DNA mismatch repair system results in insertion or deletion of single nucleotides at short repetitive DNA sequences. About 15% of sporadic and approximately 90% of hereditary nonpolyposis colorectal cancers display MSI-H. When affecting coding regions, MSI-H results in frameshift mutations and expression of corresponding frameshift peptides (FSPs). Functional tumor promoting relevance has been demonstrated for a growing number of genes frequently hit by MSI-H. Contrary, immune reactions against FSPs are involved in the immune surveillance of MSI-H cancers. Here, we provide conclusive data that the (-1) frame of U79260(FTO) encodes an HLA-A0201-restricted cytotoxic T cell epitope (FSP11; TLSPGWSAV). T cells specific for FSP11 efficiently recognized HLA-A0201((pos)) tumor cells harboring the mutated reading frame. Considering the exceptionally high mutation rate of U79260(FTO) in MSI-H colorectal carcinoma (81.8%), this recommends that FSP11 be a component of future vaccines.