Skip to Content
Merck
Fast Red TR Salt hemi(zinc chloride) salt

Fast Red TR Salt hemi(zinc chloride) salt

Fast Red TR Salt hemi(zinc chloride) salt
Synonym(s):
4-Chloro-2-methylbenzenediazonium salt, Azoic Diazo No. 11
Empirical Formula (Hill Notation):
C7H6Cl2N2 · 0.5 ZnCl2
CAS Number:
Molecular Weight:
257.19

Products

Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing