Ammonium acetate
Synonym(s):
Ammonium acetate, OmniPur® Ammonium Acetate
Linear Formula:
CH3CO2NH4
CAS Number:
Molecular Weight:
77.08
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing