Skip to Content
Merck
Seleno-L-methionine ≥98% (TLC), powder

Seleno-L-methionine

Seleno-L-methionine ≥98% (TLC), powder
Synonym(s):
(S)-2-Amino-4-(methylseleno)butyric acid, L-Selenomethionine
Linear Formula:
CH3SeCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
196.11

Products

Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer