N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride
Synonym(s):
N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDAC, EDC, EDC hydrochloride, WSC hydrochloride, EDAC, Hydrochloride, N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride
Empirical Formula (Hill Notation):
C8H17N3 · HCl
Products
Brand
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing