Skip to Content
Merck
1,4-Dichloro-2-butene (cis+trans) technical grade, 90%

1,4-Dichloro-2-butene (cis+trans)

1,4-Dichloro-2-butene (cis+trans) technical grade, 90%
Linear Formula:
ClCH2CH=CHCH2Cl
CAS Number:
Molecular Weight:
125.00

Products

Brand
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing