N,N,N′,N′-Tetramethylethylenediamine
Synonym(s):
1,2-Bis(dimethylamino)ethane, TEMED, TMEDA, N,N,N′,N′-Tetramethyl ethylenediamine (Temed), N,N,N′,N′-Tetramethyl ethylenediamine
Linear Formula:
(CH3)2NCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
116.20
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing