Merck
  • Netrin-1 is associated with macrophage infiltration and polarization in human epicardial adipose tissue in coronary artery disease.
Netrin-1 is associated with macrophage infiltration and polarization in human epicardial adipose tissue in coronary artery disease.
Journal of cardiology (2016-10-12)
Kadri Murat Gurses, Fusun Ozmen, Duygu Kocyigit, Nilgun Yersal, Elif Bilgic, Erkan Kaya, Cagla Zubeyde Kopru, Tolga Soyal, Suat Doganci, Lale Tokgozoglu, Petek Korkusuz
ABSTRACT
Inflammatory activity originating from the epicardial adipose tissue (EAT) may have a role in coronary artery disease (CAD) pathogenesis. The relationship between macrophage infiltration, polarization in the EAT, and netrin-1 gene expression was investigated. Macrophage infiltration and polarization were examined by immunohistochemical methods and expression levels of netrin-1, Unc5b, and cytokines related with M1-macrophage subtype (IL-12 and IL-18) were determined by quantitative polymerase chain reaction in subcutaneous and epicardial adipose tissue obtained from patients undergoing coronary artery bypass grafting and non-coronary cardiac surgery. CAD patients had higher CD68+ (p=0.005) and CD11c+ (p<0.001) macrophage count in EAT when compared to the controls. CD11c+/CD206+ macrophage ratio, which reflects dominancy of M1-macrophage phenotype, was significantly increased in EAT of CAD patients when compared to that of the controls (p=0.008). CAD patients had significantly higher netrin-1, Unc5b, and IL-18 gene expression in the EAT when compared to the control group (p<0.001, p<0.001, and p=0.006 respectively). Increased macrophage infiltration and polarization were associated with higher netrin-1, Unc5b, and IL-12 gene expression in EAT (p<0.05). Findings suggest a link between enhanced netrin-1 expression in EAT and macrophage infiltration and polarization in patients with CAD.