Merck
  • Potential of porphyrins as chromogenic reagents for determining metals in capillary electrophoresis.
Potential of porphyrins as chromogenic reagents for determining metals in capillary electrophoresis.
Journal of chromatography. A (2009-07-22)
S Almeda, H E Gandolfi, L Arce, M Valcárcel
ABSTRACT
Although capillary electrophoresis (CE) with photometric detection is a well-established technique for the determination of various inorganic ions, its limited sensitivity has hindered greater development in this area. In this work, we used a mixture of metals consisting of Co(II), Ni(II), Zn(II) and Mn(II) to demonstrate that the sensitivity of CE with ultraviolet-visible (UV-vis) detection can be improved by using chromogenic reagents such as porphyrins. To this end, the metals were reacted with 5,10,15,20-tetrakis(4-sulphophenyl)-porphine dodecahydrate (TPPS(4)) to obtain their respective porphyrinato complexes, which were then separated by CE with a citrate buffer and detected at 410 nm. The ensuing electrophoretic method has a limit of detection (LOD) of 3 x 10(-6) M (180 microg L(-1)) for Co(II), 2 x 10(-10) M (0.012 microg L(-1)) for Ni(II), 4 x 10(-6) M (260 microg L(-1)) for Zn(II) and 4 x 10(-9) M (0.219 microg L(-1)) for Mn(II). The method is a highly promising choice for the ultratrace determination of Ni(II) and Mn(II).
Product Description
Supelco
4,4′,4″,4″′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid), for spectrophotometric det. of transition metals, ≥95.0%