Merck
  • Innate immune receptor NOD2 promotes vascular inflammation and formation of lipid-rich necrotic cores in hypercholesterolemic mice.
Innate immune receptor NOD2 promotes vascular inflammation and formation of lipid-rich necrotic cores in hypercholesterolemic mice.
European journal of immunology (2014-07-22)
Maria E Johansson, Xiao-Ying Zhang, Kristina Edfeldt, Anna M Lundberg, Malin C Levin, Jan Borén, Wei Li, Xi-Ming Yuan, Lasse Folkersen, Per Eriksson, Ulf Hedin, Hann Low, Dmitri Sviridov, Francisco J Rios, Göran K Hansson, Zhong-Qun Yan
ABSTRACT
Atherosclerosis is an inflammatory disease associated with the activation of innate immune TLRs and nucleotide-binding oligomerization domain-containing protein (NOD)-like receptor pathways. However, the function of most innate immune receptors in atherosclerosis remains unclear. Here, we show that NOD2 is a crucial innate immune receptor influencing vascular inflammation and atherosclerosis severity. 10-week stimulation with muramyl dipeptide (MDP), the NOD2 cognate ligand, aggravated atherosclerosis, as indicated by the augmented lesion burden, increased vascular inflammation and enlarged lipid-rich necrotic cores in Ldlr(-/-) mice. Myeloid-specific ablation of NOD2, but not its downstream kinase, receptor-interacting serine/threonine-protein kinase 2, restrained the expansion of the lipid-rich necrotic core in Ldlr(-/-) chimeric mice. In vitro stimulation of macrophages with MDP enhanced the uptake of oxidized low-density lipoprotein and impaired cholesterol efflux in concordance with upregulation of scavenger receptor A1/2 and downregulation of ATP-binding cassette transporter A1. Ex vivo stimulation of human carotid plaques with MDP led to increased activation of inflammatory signaling pathways p38 MAPK and NF-κB-mediated release of proinflammatory cytokines. Altogether, this study suggests that NOD2 contributes to the expansion of the lipid-rich necrotic core and promotes vascular inflammation in atherosclerosis.
Sigma-Aldrich
SP600125, ≥98% (HPLC)