Merck
  • Multiplex Aptamer Discovery through Apta-Seq and Its Application to ATP Aptamers Derived from Human-Genomic SELEX.
Multiplex Aptamer Discovery through Apta-Seq and Its Application to ATP Aptamers Derived from Human-Genomic SELEX.
ACS chemical biology (2017-07-01)
Michael M Abdelsayed, Bao T Ho, Michael M K Vu, Julio Polanco, Robert C Spitale, Andrej Lupták
ABSTRACT
Laboratory-evolved RNAs bind a wide variety of targets and serve highly diverse functions, including as diagnostic and therapeutic aptamers. The majority of aptamers have been identified using in vitro selection (SELEX), a molecular evolution technique based on selecting target-binding RNAs from highly diverse pools through serial rounds of enrichment and amplification. In vitro selection typically yields multiple distinct motifs of highly variable abundance and target-binding affinities. The discovery of new aptamers is often limited by the difficulty of characterizing the selected motifs, because testing of individual sequences tends to be a tedious process. To facilitate the discovery of new aptamers within in vitro selected pools, we developed Apta-Seq, a multiplex analysis based on quantitative, ligand-dependent 2' acylation of solvent-accessible regions of the selected RNA pools, followed by reverse transcription (SHAPE) and deep sequencing. The method reveals, in a single sequencing experiment, the identity, structural features, and target dissociation constants for aptamers present in the selected pool. Application of Apta-Seq to a human genomic pool enriched for ATP-binding RNAs yielded three new aptamers, which together with previously identified human aptamers suggest that ligand-binding RNAs may be common in mammals.
Sigma-Aldrich
2-(Azidomethyl)nicotinic acid imidazolide, ≥95%