Merck
  • Facile Synthesis of Well-Dispersed Ni2P on N-Doped Nanomesh Carbon Matrix as a High-Efficiency Electrocatalyst for Alkaline Hydrogen Evolution Reaction.
Facile Synthesis of Well-Dispersed Ni2P on N-Doped Nanomesh Carbon Matrix as a High-Efficiency Electrocatalyst for Alkaline Hydrogen Evolution Reaction.
Nanomaterials (Basel, Switzerland) (2019-07-20)
Fan Yang, Shuo Huang, Bing Zhang, Liqiang Hou, Yi Ding, Weijie Bao, Chunming Xu, Wang Yang, Yongfeng Li
ABSTRACT
The development of non-noble metal hydrogen evolution catalysts that can replace Pt is crucial for efficient hydrogen production. Herein, we develop a type of well-dispersed Ni2P on N-doped nanomesh carbon (NC) electrocatalyst by a facile pyrolysis method, which shows excellent hydrogen evolution reaction (HER) catalytic performance. It is rather remarkable that the overpotential of Ni2P/NC prepared under optimal proportion is 108 mV at 10 mA·cm-2 current density in 1 M KOH solution with the tafel slope of 67.3 mV·dec-1, the catalytic activity has no significant attenuation after 1000 cycles of cyclic voltammetry (CV)method. The hydrogen evolution performance of the electrocatalytic is better than most similar catalysts in alkaline media. The unique mesh structure of the carbon component in the catalyst facilitates the exposure of the active site and reduces the impedance, which improves the efficiency of electron transport as well as ensuring the stability of the hydrogen evolution reaction. In addition, we prove that nitrogen doping and pore structure are also important factors affecting catalytic activity by control experiments. Our results show that N-doped nanomesh carbon, as an efficient support, combined with Ni2P nanoparticles is of great significance for the development of efficient hydrogen evolution electrodes.