Tryptone enzymatic digest from casein
Synonym(s):
Peptone from casein
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
pH Value
Physical Form
Tissue
Chemical Composition
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing