Skip to Content
Merck
Methacrylic acid - methylmethacrylate copolymer (1:2) European Pharmacopoeia (EP) Reference Standard

Methacrylic acid copolymer type B

Methacrylic acid - methylmethacrylate copolymer (1:2) European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Poly(methyl methacrylate-co-methacrylic acid), Poly(methacrylic acid-co-methyl methacrylate)
CAS Number:

Products

Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade