Skip to Content
Merck
Insulin human ≥95% (HPLC), semisynthetic, powder, non-sterile

Insulin human

Insulin human ≥95% (HPLC), semisynthetic, powder, non-sterile
Synonym(s):
Insulin human, Insulin sodium salt human, Insulin, Zinc, Human, Recombinant, P. pastoris, Insulin zinc salt human, Zinc insulin crystals human
Empirical Formula (Hill Notation):
C1542H2298N390O462S36Zn2
CAS Number:
Molecular Weight:
34976.20

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Recombinant Host
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Technique