(±)-α-Tocopherol
Synonym(s):
DL-all-rac-α-Tocopherol, Vitamin E, (±)-α-Tocopherol, all-rac-α-Tocopherol
Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing