Skip to Content
Merck
Benzoic acid purified by sublimation, ≥99%

Benzoic acid

Benzoic acid purified by sublimation, ≥99%
Synonym(s):
Benzoic acid
Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
Special Grade
Technique