Merck
  • An atmospheric pressure chemical ionisation liquid chromatographic-tandem mass spectrometry method for the analysis of benzodiazepines in urine.
An atmospheric pressure chemical ionisation liquid chromatographic-tandem mass spectrometry method for the analysis of benzodiazepines in urine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-09-11)
S Dunlop, K Hayes, P Leavy, D Cusack, R Maguire
ABSTRACT
The objective of this work was to establish an analytical method for the analysis of 7 Benzodiazepines (diazepam, oxazepam, temazepam, nordiazepam, desalkylflurazepam, alprazolam and α-hydroxyalprazolam) in urine specimens taken from drivers suspected of driving under the influence of drugs. The specimen, calibrator and control preparation involved hydrolysis of conjugated benzodiazepines using β-glucuronidase in sodium acetate buffer, with incubation at 60°C for 2h. Specimens were then centrifuged, before being diluted 1 in 5 (total dilution 1 in 10), with 10% acetonitrile in water. Specimens were analysed using a Shimadzu Prominence UPLC coupled to an AB Sciex 4000 QTrap LC-MS-MS. The chromatographic column was a Shim-pack XR ODS 2.2μm. 3.0×50mm column and the mobile phase was a binary gradient system comprising of mobile phase A which was an ammonium formate/formic acid buffer dissolved in water and mobile phase B which was an ammonium formate/formic acid buffer dissolved in Acetonitrile. APCI was selected as the ionisation technique and the MS was operated in MRM mode, monitoring 2 transitions per analyte. The validation of the method is described. The method was found to be linear, accurate and precise (within day and between day) for diazepam, oxazepam, temazepam, nordiazepam, desalkylflurazepam, alprazolam and α-hydroxyalprazolam. The results of 480 cases are reviewed and show that alprazolam use was found in 35% of cases. Use of benzodiazepines resulting in oxazepam, nordiazepam or temazepam were found ca. 70% of cases analysed.
Product Description
Supelco
Norfludiazepam, analytical standard