Merck
  • Kinetic effect of oestrogen on secretion of prostaglandins E2 and F2α in bovine oviduct epithelial cells.
Kinetic effect of oestrogen on secretion of prostaglandins E2 and F2α in bovine oviduct epithelial cells.
Reproduction, fertility, and development (2017-04-27)
Zhiheng Dong, Nan Zhang, Wei Mao, Bo Liu, Na Huang, Peifeng Li, Changyou Li, Jinshan Cao
ABSTRACT
This study aimed to investigate the effect of oestrogen on prostaglandin E2 (PGE