Merck
  • LncRNA-HOTAIR promotes TNF-α production in cardiomyocytes of LPS-induced sepsis mice by activating NF-κB pathway.
LncRNA-HOTAIR promotes TNF-α production in cardiomyocytes of LPS-induced sepsis mice by activating NF-κB pathway.
Biochemical and biophysical research communications (2016-01-26)
Hongwei Wu, Jinfeng Liu, Wei Li, Gang Liu, Zhenguang Li
ABSTRACT
Mounting studies have illustrated an important role of HOTAIR in cancer progress, but few studies have reported its function in cardiac disease, including cardiac-associated sepsis. This study aimed to investigate the function of HOTAIR in sepsis, involving its association with the level of tumor necrosis factor-alpha (TNF-α), an important inducer of myocardial dysfunction during LPS-induced sepsis. Sepsis mice model was established by LPS administration, and myocardial dysfunction was evaluated with hemodynamic parameters. HOTAIR expression in isolated cardiomyocytes and TNF-α production in the circulation were detected, as well as the protein levels of phosphorylated p65. HL-1 cells were subjected to LPS treatment in vitro for functional studies, including luciferase report assays for NF-κB activity. HOTAIR expression was significantly upregulated in cardiomyocytes from sepsis mice, in line with increased TNF-α production and p65 phosphorylation, while similar results were also observed in LPS treated HL-1 cells, which was then reversed by HOTAIR interference. Functional studies demonstrated that HOTAIR showed positive regulation on p65 phosphorylation and NF-κB activation, while HOTAIR-induced TNF-α production was repressed by NF-κB inhibitor. Further in vivo studies confirmed that HOTAIR silence can improve cardiac function of sepsis mice, and markedly decreased TNF-α production in the circulation. HOTAIR upregulation in cardiomyocytes of LPS-induced sepsis mice promoted TNF-α production in the circulation by activating NF-κB, involving the phosphorylation of NF-κB p65 subunit. Moreover, HOTAIR silence preserved cardiac function of sepsis mice during LPS-induced sepsis.
Sigma-Aldrich
Claycomb Medium, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Germane, electronic grade, ≥99.997%