Merck
  • Activity-dependent neuroprotector homeobox protein: A candidate protein identified in serum as diagnostic biomarker for Alzheimer's disease.
Activity-dependent neuroprotector homeobox protein: A candidate protein identified in serum as diagnostic biomarker for Alzheimer's disease.
Journal of proteomics (2012-05-05)
Ming-Hui Yang, Yuan-Han Yang, Chi-Yu Lu, Shiang-Bin Jong, Li-Jhen Chen, Yu-Fen Lin, Shyh-Jong Wu, Pei-Yu Chu, Tze-Wen Chung, Yu-Chang Tyan
ABSTRACT
Alzheimer's disease (AD) is the most common cause of dementia of late life. To enhance our understanding of AD proteome, the serum proteins were analyzed using two-dimensional gel electrophoresis (2DE) combined with nano-high performance liquid chromatography electrospray ionization tandem mass spectrometry (nano-HPLC-ESI-MS/MS) followed by peptide fragmentation patterning. In this study, six protein spots with differential expression were identified. Five up-regulated proteins were identified as actin, apolipoprotein A-IV (Apo A-IV), inter-alpha-trypsin inhibitor heavy chain H4 (ITIH4), alpha-1-antitrypsin (AAT), and antithrombin-III (AT-III); one protein, activity-dependent neuroprotector homeobox protein (ADNP) was down-regulated in AD patients. These proteins with differential expression in the serum may serve as potential indicators of AD. Our results suggested that ADNP may play an important role in slowing the progression of clinical symptoms of AD.