Merck
  • G-quadruplex-mediated regulation of telomere binding protein POT1 gene expression.
G-quadruplex-mediated regulation of telomere binding protein POT1 gene expression.
Biochimica et biophysica acta (2014-03-19)
Qingqing He, Ping Zeng, Jia-Heng Tan, Tian-Miao Ou, Lian-Quan Gu, Zhi-Shu Huang, Ding Li
ABSTRACT
Telomere is protected by its G-quadruplex, T-loop structure, telomerase, and binding protein complex. Protein POT1 (protection of telomeres 1) is one subunit of telomere binding protein complex Shelterin. POT1 acts as a regulator of telomerase-dependent telomere length, and it can help telomere to form D-loop structure to stabilize telomere. POT1 protects telomere ends from ATR-dependent DNA damage response as well. Extensive methods were used, including CD, EMSA, ITC, PCR stop assay, luciferase reporter assay, quantitative real-time PCR, Western blot, chromatin immunoprecipitation (Ch-IP), cloning, expression and purification of proteins. We found a new G-rich 30-base-pair long sequence (P-pot1 G18) located from -165 to -136 base pairs upstream of the translation starting site of protein POT1. This sequence in the promoter region of pot1 gene formed G-quadruplex resulting in down-regulation of pot1 gene transcription. This G-rich sequence is close to a binding site "TCCC" for transcription factor hnRNP K (heterogeneous nuclear ribonucleoprotein K), and its conversion to G-quadruplex prevented the access of hnRNP K to this binding site. The binding of hnRNP K could up-regulate pot1 gene transcription. TMPyP4 (meso-tetra(N-methyl-4-pyridyl)porphine) has been widely used as G-quadruplex binding ligand, which stabilized the G-quadruplex in vitro and in cellulo, resulting in down-regulation of pot1 gene transcription. This G-quadruplex might become a potentially new drug target for antitumor agents. Our results first demonstrated that G-quadruplex formation can affect the binding of transcription factor to its nearby binding site, and thus making additional influence to gene transcription.
Sigma-Aldrich
5,10,15,20-Tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate), Dye content 90 %