Merck
  • Evaluation of the in-vitro activity of furopenem (SY5555 or SUN5555) against respiratory tract pathogens and beta-lactamase producing bacteria.
Evaluation of the in-vitro activity of furopenem (SY5555 or SUN5555) against respiratory tract pathogens and beta-lactamase producing bacteria.
The Journal of antimicrobial chemotherapy (1995-04-01)
M G Cormican, R N Jones
ABSTRACT
Furopenem is a novel orally active penem. In this study, furopenem was highly active in vitro against Streptococcus pneumoniae (MIC90 0.03 mg/L), Haemophilus influenzae (MIC90, 2 mg/L), and Moraxella catarrhalls (MIC90, 0.5 mg/L). Its activity was not reduced by a variety of beta-lactamase enzymes, however beta -lactam resistance by other mechanisms was associated with higher MICs.
Sigma-Aldrich
Faropenem sodium hydrate, ≥98% (HPLC)