Merck
  • [123I]Iodooctyl fenbufen amide as a SPECT tracer for imaging tumors that over-express COX enzymes.
[123I]Iodooctyl fenbufen amide as a SPECT tracer for imaging tumors that over-express COX enzymes.
Biomaterials (2013-02-07)
Ho-Lien Huang, Chun-Nan Yeh, Wei-Yuan Lee, Ying-Cheng Huang, Kang-Wei Chang, Kun-Ju Lin, Shu-Fan Tien, Wen-Chin Su, Ching-Hsiuan Yang, Jenn-Tzong Chen, Wuu-Jyh Lin, Shio-Shio Fan, Chung-Shan Yu
ABSTRACT
This study is concerned with the development of an agent for single photon emission computer tomography (SPECT) for imaging inflammation and tumor progression. [(123)I]Iodooctyl fenbufen amide ([(123)I]IOFA) was prepared from the precursor N-octyl-4-oxo-4-(4'-(trimethylstannyl)biphenyl-4-yl)butanamide with a radiochemical yield of 15%, specific activity of 37 GBq/μmol, and radiochemical purity of 95%. Analysis of the binding of [(123)I]IOFA to COX-1 and COX-2 enzymes by using HPLC and a gel filtration column showed a selectivity ratio of 1:1.3. An assay for the competitive inhibition of substrate transfer showed that IOFA exhibited a comparable IC(50) value compared to fenbufen. In the normal rat liver, a lower level and homogeneous pattern of [(123)I]IOFA radioactivity was observed by SPECT. In contrast, in the rat liver with thioacetamide-induced cholangiocarcinoma, a higher uptake and heterogeneous pattern of [(123)I]IOFA radioactivity was seen as hot spots in tumor lesions by SPECT imaging. Importantly, elevated COX-1 and COX-2 expressions from immunostaining were found in the bile ducts of tumor rats but not of normal rats. Therefore, [(123)I]IOFA was found to exhibit the potential for imaging tumors that over-express COX.
Product Description
Supelco
Fenbufen, analytical standard