Merck
  • The kinetic and structural characterization of the reaction of nafamostat with bovine pancreatic trypsin.
The kinetic and structural characterization of the reaction of nafamostat with bovine pancreatic trypsin.
Thrombosis research (2000-07-19)
M K Ramjee, I M Henderson, S B McLoughlin, A Padova
ABSTRACT
Nafamostat mesilate (FUT-175), a synthetic serine protease inhibitor, is active against a number of the serine proteases involved in coagulation. This has been proposed as the basis of its anticoagulant activity. We investigated the reaction of Nafamostat with bovine pancreatic trypsin as a model system. It was shown to act as a time-dependent competitive inhibitor, and the inhibition constants for the binding of Nafamostat to trypsin (i.e., Ki) and the overall inhibition constants (i.e., Ki*) were calculated to be 11.5 microM and 0.4+/-0.14 nM, respectively. The second-order rate constant for the reaction was 4.5+/-0.19x10(5) M(-1)s(-1), and the product released following the acylation step, 6-amidino2-naphthol, showed mixed-type inhibition. The competitive (Kic) and uncompetitive (Kiu) inhibition constants were 14.7 microM and 19.5 microM, respectively. Formation of the acyl-enzyme intermediate was dissected into at least two steps, with rates of 0.9 s(-1) and 195 s(-1). The deacylation step was relatively much slower (3.2+/-0.19x10(-5) s(-1), enabling the mass spectroscopic analysis of the acyl-enzyme intermediate, which confirmed the covalent attachment of 4-guanidinobenzoic acid to trypsin. The product of the deacylation step, 4-guanidinobenzoic acid, showed no inhibition up to a concentration of 200 microM. These data strongly suggest that while Nafamostat is a potent inhibitor of trypsin, it is actually an extremely poor substrate, and that apparent inhibition is due to the competitive formation of a very stable acyl-enzyme intermediate, analogous to some other active site titrants.
Sigma-Aldrich
4-Guanidinobenzoic acid hydrochloride, 99%