Merck
  • P-glycoprotein (ABCB1) and breast cancer resistance protein (ABCG2) localize in the microvessels forming the blood-tumor barrier in ependymomas.
P-glycoprotein (ABCB1) and breast cancer resistance protein (ABCG2) localize in the microvessels forming the blood-tumor barrier in ependymomas.
Brain pathology (Zurich, Switzerland) (2010-04-22)
Carole Ginguené, Jacques Champier, Said Maallem, Nathalie Strazielle, Anne Jouvet, Michelle Fèvre-Montange, Jean-François Ghersi-Egea
ABSTRACT
Ependymomas are neuroepithelial tumors that arise from the ependymal layer bordering the cerebral ventricles and spinal canal. Intracranial ependymoma represents a major encephalic tumor in children, while spinal ependymoma develops more frequently in adults. To understand the pharmacoresistance that characterizes this tumoral entity, we analyzed the level of expression and localization of three major efflux transport proteins with a multidrug resistance function, P-glycoprotein, multidrug resistance-related protein 1 (MRP1) and breast cancer resistance protein (BCRP), in a series of 25 ependymomas from both children and adults. Real-time-PCR analysis showed that all three genes were expressed in all tumors, with no apparent correlation between the level of expression and either age or tumor grade. The MRP1 transcript was expressed at a significantly higher level in spinal tumors than in intracranial tumors. The expression of the proteins corresponding to these genes was confirmed by Western blot analysis. In an immunohistochemical study, P-glycoprotein and BCRP were shown to be associated with the tumoral vessels, where they presented a luminal localization, a prerequisite for their efflux drug activity into the blood. These data indicate that a biochemical, transporter-dependent blood-tumor barrier may exist in ependymomas, which may reduce the tumoral bioavailability of lipophilic and amphiphilic anticancer drugs.
Sigma-Aldrich
Anti-P-Glycoprotein Mouse mAb (C219), liquid, clone C219, Calbiochem®