Merck
  • Combined TCR Repertoire Profiles and Blood Cell Phenotypes Predict Melanoma Patient Response to Personalized Neoantigen Therapy plus Anti-PD-1.
Combined TCR Repertoire Profiles and Blood Cell Phenotypes Predict Melanoma Patient Response to Personalized Neoantigen Therapy plus Anti-PD-1.
Cell reports. Medicine (2020-12-10)
Asaf Poran, Julian Scherer, Meghan E Bushway, Rana Besada, Kristen N Balogh, Amy Wanamaker, Reid G Williams, Jasmina Prabhakara, Patrick A Ott, Siwen Hu-Lieskovan, Zakaria S Khondker, Richard B Gaynor, Michael S Rooney, Lakshmi Srinivasan
ABSTRACT
T cells use highly diverse receptors (TCRs) to identify tumor cells presenting neoantigens arising from genetic mutations and establish anti-tumor activity. Immunotherapy harnessing neoantigen-specific T cells to target tumors has emerged as a promising clinical approach. To assess whether a comprehensive peripheral mononuclear blood cell analysis predicts responses to a personalized neoantigen cancer vaccine combined with anti-PD-1 therapy, we characterize the TCR repertoires and T and B cell frequencies in 21 patients with metastatic melanoma who received this regimen. TCR-α/β-chain sequencing reveals that prolonged progression-free survival (PFS) is strongly associated with increased clonal baseline TCR repertoires and longitudinal repertoire stability. Furthermore, the frequencies of antigen-experienced T and B cells in the peripheral blood correlate with repertoire characteristics. Analysis of these baseline immune features enables prediction of PFS following treatment. This method offers a pragmatic clinical approach to assess patients' immune state and to direct therapeutic decision making.
Product Description
Millipore
Benzonase® Nuclease, Purity > 90%