Merck
  • Lateral hypothalamic NMDA receptor subunits NR2A and/or NR2B mediate eating: immunochemical/behavioral evidence.
Lateral hypothalamic NMDA receptor subunits NR2A and/or NR2B mediate eating: immunochemical/behavioral evidence.
The American journal of physiology (1999-03-10)
A M Khan, M C Currás, J Dao, F A Jamal, C A Turkowski, R K Goel, E R Gillard, S D Wolfsohn, B G Stanley
ABSTRACT
Cells within the lateral hypothalamic area (LHA) are important in eating control. Glutamate or its analogs, kainic acid (KA) and N-methyl-D-aspartate (NMDA), elicit intense eating when microinjected there, and, conversely, LHA-administered NMDA receptor antagonists suppress deprivation- and NMDA-elicited eating. The subunit composition of LHA NMDA receptors (NMDA-Rs) mediating feeding, however, has not yet been determined. Identifying this is important, because distinct second messengers/modulators may be activated by NMDA-Rs with differing compositions. To begin to address this, we detected LHA NR2A and NR2B subunits by immunoblotting and NR2B subunits by immunohistochemistry using subunit-specific antibodies. To help determine whether NMDA-Rs mediating feeding might contain these subunits, we conducted behavioral studies using LHA-administered ifenprodil, an antagonist selective for NR2A- and/or NR2B-containing NMDA-Rs at the doses we used (0.001-100 nmol). Ifenprodil maximally suppressed NMDA- and deprivation-elicited feeding by 63 and 39%, respectively, but failed to suppress KA-elicited eating, suggesting its actions were behaviorally specific. Collectively, these results suggest that LHA NMDA-Rs, some of which contribute to feeding control, are composed of NR2A and/or NR2B subunits, and implicate NR2A- and/or NR2B-linked signal transduction in feeding behavior.