1,4-Dioxane
Synonym(s):
Diethylene oxide, Dioxane, 1,4-Dioxane solution, 1,4-Dioxane
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing