Diltiazem Hydrochloride
Synonym(s):
(+)-cis-Diltiazem hydrochloride, (2S,3S)-(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, CRD-401, Diltiazem, Hydrochloride
Empirical Formula (Hill Notation):
C22H26N2O4S · HCl
CAS Number:
Molecular Weight:
450.98
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing