Skip to Content
Merck
Phenytoin for system suitability European Pharmacopoeia (EP) Reference Standard

Phenytoin

Phenytoin for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
5,5-Diphenylhydantoin, 5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin
Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique