Skip to Content
Merck
Nitrite ion standard solution 0.1 M NO2-, for ion-selective electrodes

Nitrite Standard for IC

Nitrite ion standard solution 0.1 M NO2-, for ion-selective electrodes
Synonym(s):
Sodium nitrite solution
Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00

Products

Brand
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique