Skip to Content
Merck
Hydrogen peroxide solution 31% Ultrapur

Hydrogen peroxide solution

Hydrogen peroxide solution 31% Ultrapur
Synonym(s):
Hydrogen peroxide solution
Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01

Products

Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Markush Class
Markush Group
Microorganism Suitability
Reagent Type
Special Grade
Technique