Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(ethyleneimine) solution
Synonym(s):
Ethyleneimine polymer solution, PEI
Linear Formula:
H(NHCH
2
CH
2
)
m
NH
2
CAS Number:
9002-98-6
Products
Brand
Millipore (1)
Sigma-Aldrich (5)
Supelco (1)
Biological Source
Color
Physical Form
Format
Tissue
Quality Segment
Application
Polymer Composition
Polymer Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
181978
Poly(ethyleneimine) solution,
average M
n
~60,000 by GPC, average M
w
~750,000 by LS, 50 wt. % in H
2
O
Expand
View Pricing
P3143
Poly(ethyleneimine) solution,
analytical standard, 50 % (w/v) in H
2
O
Expand
View Pricing
408700
Poly(ethyleneimine) solution,
average M
w
2,000 by LS, 50 wt. % in H
2
O
Expand
View Pricing
482595
Poly(ethyleneimine) solution,
average M
n
~1,200, average M
w
~1300 by LS, 50 wt. % in H
2
O
Expand
View Pricing
913375
Poly(ethyleneimine) solution,
2mg/mL aqueous solution, suitable for biomedical research
Expand
View Pricing
03880
Poly(ethyleneimine) solution,
~50% in H
2
O
Expand
View Pricing
71412
Poly(ethyleneimine) solution,
A ready-to-use, stabilized lysozyme solution
Expand
View Pricing