Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium 1,2-naphthoquinone-4-sulfonate
Synonym(s):
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 3,4-Dihydro-3,4-dioxo-1-naphthalenesulfonic acid sodium salt, Folin’s reagent
Empirical Formula (Hill Notation):
C
10
H
5
NaO
5
S
CAS Number:
521-24-4
Molecular Weight:
260.20
EC Number:
208-308-9
Products
Brand
Supelco (1)
Formula Weight
Physical Form
Purity
Analyte Suitability
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
70382
Sodium 1,2-naphthoquinone-4-sulfonate,
for spectrophotometric det. of isonicotic hydrazide, amines and amino acids, ≥97.0% (T)
Expand
View Pricing