Skip to Content
Merck
(S)-(+)-2-Aminoheptane 99%

(S)-(+)-2-Aminoheptane

(S)-(+)-2-Aminoheptane 99%
Linear Formula:
CH3(CH2)4CH(NH2)CH3
CAS Number:
Molecular Weight:
115.22

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing