Skip to Content
Merck
(R)-(−)-Mevalonolactone ≥90.0% (GC)

(R)-(−)-Mevalonolactone

(R)-(−)-Mevalonolactone ≥90.0% (GC)
Synonym(s):
(R)-3-Hydroxy-3-methyl-δ-valerolactone, (R)-Mevalolactone, D-Mevalonic acid lactone
Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing