Skip to Content
Merck
(R)-(−)-2-Butanol 99%

(R)-(−)-2-Butanol

(R)-(−)-2-Butanol 99%
Synonym(s):
(R)-(−)-sec-Butyl alcohol
Linear Formula:
C2H5CH(OH)CH3
CAS Number:
Molecular Weight:
74.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing