![Neocuproine hemihydrate for spectrophotometric det. of Cu, ≥99.0%](/deepweb/assets/sigmaaldrich/product/structures/227/899/ad49d4bf-de7a-4b38-a8ce-2b3bd1c9a486/640/ad49d4bf-de7a-4b38-a8ce-2b3bd1c9a486.png)
Neocuproine hemihydrate
![Neocuproine hemihydrate for spectrophotometric det. of Cu, ≥99.0%](/deepweb/assets/sigmaaldrich/product/structures/227/899/ad49d4bf-de7a-4b38-a8ce-2b3bd1c9a486/640/ad49d4bf-de7a-4b38-a8ce-2b3bd1c9a486.png)
Synonym(s):
2,9-Dimethyl-1,10-phenanthroline
Empirical Formula (Hill Notation):
C14H12N2 · 0.5H2O
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing