
Methyl 2-(bromomethyl)acrylate

Linear Formula:
H2C=C(CH2Br)CO2CH3
CAS Number:
Molecular Weight:
179.01
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing