Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium aluminate
Linear Formula:
MgO·Al
2
O
3
CAS Number:
12068-51-8
Molecular Weight:
142.27
EC Number:
235-100-5
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Purity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
634832
Magnesium aluminate,
single crystal substrate, <111>, 99.99% trace metals basis
Expand
View Pricing
634840
Magnesium aluminate,
single crystal substrate, <110>, 99.99% trace metals basis
Expand
View Pricing