Skip to Content
Merck
Luperox® A70S, Benzoyl peroxide 70%, remainder water

Luperox® A70S, Benzoyl peroxide

Luperox® A70S, Benzoyl peroxide 70%, remainder water
Synonym(s):
Benzoyl peroxide, Dibenzoyl peroxide
Linear Formula:
(C6H5CO)2O2
CAS Number:
Molecular Weight:
242.23

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing