Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lead(IV) acetate
Synonym(s):
Lead tetraacetate, Pb(acac)
4
Linear Formula:
Pb(CH
3
CO
2
)
4
CAS Number:
546-67-8
Molecular Weight:
443.38
EC Number:
208-908-0
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
185191
Lead(IV) acetate,
reagent grade, 95%
Expand
View Pricing