Skip to Content
Merck
Imazalil sulfate PESTANAL®, analytical standard

Imazalil sulfate

Imazalil sulfate PESTANAL®, analytical standard
Synonym(s):
1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy)ethyl]-1H-imidazole sulfate, Enilconazole sulfate
Empirical Formula (Hill Notation):
C14H14Cl2N2O · H2SO4
CAS Number:
Molecular Weight:
395.26
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing