Skip to Content
Merck
IAEA-N-1 (nitrogen isotopes in ammonium sulfate) NIST® RM 8547

IAEA-N-1 (nitrogen isotopes in ammonium sulfate)

IAEA-N-1 (nitrogen isotopes in ammonium sulfate) NIST® RM 8547
Synonym(s):
Ammonium sulfate, Mascagnite
Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Isotope
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing